Please login to your online account to display your discounted pricing

Omnifit “T” Series Bottle Caps


Easy to use and versatile range of caps requires no fittings

Manufacturer: bio chem fluidics inc  00938T3

 View more versions of this product

Catalog No. OF938T3

  • / Each
 

Description & Specifications

Specifications

Material Polytetrafluoroethylene
Diameter (Metric) Outer 54mm
Includes Three port
Height (Metric) With Closure 28.5mm
For Use With T series solvent bottle
No. per Case 1 Cs.
Neck Finish 38-430