Diba Omnifit™ T-Series Bottle Caps
Easy to use and versatile range of caps requires no fittings
$77.62 - $175.88
Specifications
For Use With (Equipment) | T series solvent bottle |
---|---|
Material | PTFE |
No. per Case | 1 Cs. |
Catalog Number | Mfr. No. | Thread Size (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thread Size (Metric) | Includes | Price | Quantity | ||||
OF945T2
|
Diba
00945T2 |
GL 45 | Two port |
Each for $77.62
|
|
||||
OF938T2F
|
Diba
00938T2F |
Built-in check valve and inlet filter, two port |
Each for $152.42
|
|
|||||
OF945T3F
|
Diba
00945T3F |
GL 45 | Built-in check valve and inlet filter, three port |
Each for $175.88
|
|
||||
OF945T4F
|
Diba
00945T4F |
GL 45 | Built-in check valve and inlet filter, four port |
Each for $149.38
|
|
||||
OF932T2
|
Diba
00932T2 |
GL 32 | Two port |
Each for $147.84
|
|
||||
OF945T3
|
Diba
00945T3 |
GL 45 | Three port |
Each for $79.30
|
|
||||
OF938T4
|
Diba
00938T4 |
Four port |
Each for $141.91
|
|
|||||
OF945T2F
|
Diba
00945T2F |
GL 45 | Built-in check valve and inlet filter, two port |
Each for $136.72
|
|
||||
OF938T3
|
Diba
00938T3 |
Three port |
Each for $131.91
|
|
|||||
OF938T2
|
Diba
00938T2 |
Two port |
Each for $138.22
|
|
|||||
OF945T4
|
Diba
00945T4 |
GL 45 | Four port |
Each for $90.32
|
|
||||
Description
Easy to use and versatile range of caps requires no fittings
- Luer ported, straight through tubing
- Simply push 1/8 in. O.D. hard wall tubing through the ports
- For 1/16 in. O.D. hard wall tubing, use OF9LA adapter supplied with cap
- Use OF9BA series adapter for softwall tubing
- Anti-twist design prevents tubing from kinking when removing cap
- Each port also accepts male Luer connectors
- With polypropylene screw cap and chemically inert solid PTFE body
Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.
Order Info
Available with Luer port or Luer port with fluorocarbon check valve and 10μm porosity PTFE inlet filter enabling air to flow into the bottle while preventing particulate contamination and preventing harmful vapors from escaping.
Specifications
T series solvent bottle | |
PTFE | |
1 Cs. |