Please login to your online account to display your discounted pricing
Omnifit “T” Series Bottle Caps
Easy to use and versatile range of caps requires no fittings
Manufacturer: bio chem fluidics inc  00938T4F
 View more versions of this product
Catalog No. OF938T4F
  • / Each
Description & Specifications
Specifications
Includes Built-in check valve and inlet filter, four port
Height (Metric) With Closure 28.5mm
Material PTFE
No. per Case 1 Cs.
For Use With (Equipment) T series solvent bottle
Diameter (Metric) Outer 54mm
Neck Finish 38-430