Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
$1,137.15 - $1,137.15
Specifications
For Use With Omnifit SolventPlus chromatography columns
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Includes Type Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Includes Type Price Quantity  
14-500-643 Bio Chem Fluidics Inc
006SCC0610AF
6.6mm Glass column assembly, frit kit, fittings kit; One adjustable, one fixed endpiece SolventPlus chromatography column Each for $1,137.15
Specifications
Description
  • Borosilicate glass and PTFE construction, with Chemraz™ O-rings and PTFE frits put only inert materials in the fluid path
  • ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system
  • PTFE endpiece makes the primary seal to glass and has even distribution pattern to give even sample application across the bed surface
  • Ready-assembled column includes fixed or adjustable endpieces (-FF, -AF or -AA configuration) and 30μm PE frits
Description & Specifications
Specifications
Omnifit SolventPlus chromatography columns