Please login to your online account to display your discounted pricing
Omnifit “Q” Series Bottle Caps
Easy operation ON/OFF valves offer a simple way to connect tubing
Manufacturer: bio chem fluidics inc  00945Q3
 View more versions of this product
Use OF9VF check valve and filter assembly to prevent harmful vapors from escaping and to stop contamination from entering the bottle.
Catalog No. OF945Q3
  • / Each
Description & Specifications
Specifications
Material PTFE
Diameter (Metric) Outer 54mm
For Use With (Equipment) Q series solvent bottle
Neck Finish GL45
Height (Metric) With Closure 55.7mm
No. per Case 1/Cs.
Includes Three port