Kinesis™ Omnifit™ Luer Adapter
Adapters fit into 1/4 in.–28 UNF flat bottom ports in most Omnifit connectors and valves
Manufacturer:  Kinesis™ 001206
 View more versions of this product
Catalog No. OF001206
Description
Description
  • Provide quick, push-on connection for softwall tubing or luer
  • Made from chemically resistant ETFE
  • Autoclavable
  • OF001206 features a thread to male Luer lock adapter with a polypropylene lock ring
  • OF002501 features male luer fittings to 1/4 in.–28 UNF thread
  • OF002502 features femal luer fittings to 1/4 in.–28 UNF thread
Specifications
Specifications
Luer Lock Fitting
Fluorinated Polymers
Documents