MilliporeSigma™ Supelco™ Omnifit™ Glass Columns
For use in chromatographic sample preparation, purification and traditional/classical flash chromatography
$237.54 - $301.12
Specifications
Description No Hardware
Type Column
For Use With (Application) Chromatographic Sample Preparation, Purification and Traditional/Classical Flash Chromatography
Material Glass
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Price Quantity  
111009862
View Documents Promotion Details
MilliporeSigma™ Supelco™
56018
10 mm 500 mm
Each for $282.38
111009968
View Documents Promotion Details
MilliporeSigma™ Supelco™
56024
25 mm 150 mm
Each for $237.54
111009646
View Documents Promotion Details
MilliporeSigma™ Supelco™
56026
25 mm 500 mm
Each for $301.12
Description
Description
Specifications
Specifications
No Hardware
Column
Chromatographic Sample Preparation, Purification and Traditional/Classical Flash Chromatography
Glass
SDS
missing translation for 'documents'
missing translation for 'documents'
Omnifit is a registered trademark of Diba Industries