missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Omni International Omni Tip™ Probes

Clear to ensure that sample is not lost in the probe

Manufacturer:  Omni International 34750

 View more versions of this product

Catalog No. 15-340-105



Description

Description

Disposable to eliminate cross-contamination. May also be reused and autoclaved up to seven times.

  • Slots, instead of windows, prevent sample from being trapped
  • Probes fit tubes from 1.5 to 50mL
  • Patented

  • Soft tissue probes utilize flexible tines for gentler processing
  • Hard tissue probes have saw teeth for more aggressive processing

Specifications

Specifications

4.33 in.
0.25 to 30mL
0.7cm
Ideal for processing liquids, soft tissues and samples that require gentle processing
Plastic
11cm
0.27 in.
Omni Tip Soft Tissue Probe
Omni TH, THQ, GLH, PDH and Omni prep 1.5 to 50mL tube homogenizer
Documents