Omni International Omni Prep and Omni THQ™ Stainless Steel Homogenizer Probes
Ideal for larger sample volumes, larger initial sample tissue sizes, and tougher tissue types
Manufacturer:  Omni International B5075
 View more versions of this product
Catalog No. 10-988-000
Specifications Description & Specifications
Specifications
2.95 in.
0.2 to 5mL
0.5cm
Ideal for liquid/liquid, soft tissue and microtube processing
Stainless Steel
7.5cm
0.2 in.
Probe
Omni Prep and Omni THQ Homogenizers