Please login to your online account to display your discounted pricing

Omni International Omni Mixer Fine Saw Tooth Generator Probe

$2,057.53 - $2,057.53

Specifications

Contains PTFE upper and lower bearing
Type Fine Saw Tooth Generator Probe
Material Stainless Steel
For Use With (Equipment) Omni Mixer
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

15-340-135

 
omni international inc
G795ST
7 x 95mm Each for $2,057.53
Description & Specifications

Specifications

Contains PTFE upper and lower bearing
Type Fine Saw Tooth Generator Probe
Material Stainless Steel
For Use With (Equipment) Omni Mixer