Please login to your online account to display your discounted pricing

Olopatadine hydrochloride, Tocris™

H1 antagonist; ocular antiallergic agent

$76.90 - $293.30

Chemical Identifiers

Chemical Name or Material Olopatadine hydrochloride
Assay Percent Range >99%
Molecular Formula C21H23NO3·HCl
Formula Weight 373.87
Synonym (Z)-11-[3-(Dimethylamino)propylidene]-6, 11-dihydro-dibenz[b, e]oxepin-2-acetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

424110

 
r & d systems
RDS424110
10mg Each for $76.90

424150

 
r & d systems
RDS424150
50mg Each for $293.30
Description & Specifications

Specifications

Formula Weight 373.87
Quantity 10mg
Chemical Name or Material Olopatadine hydrochloride
Assay Percent Range >99%
Molecular Formula C21H23NO3·HCl
CAS 140462-76-6
Synonym (Z)-11-[3-(Dimethylamino)propylidene]-6, 11-dihydro-dibenz[b, e]oxepin-2-acetic acid hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature