Please login to your online account to display your discounted pricing

Olomoucine, Tocris


Cyclin-dependent kinase inhibitor

Quantity 5mg

Manufacturer: r & d systems  12845

 View more versions of this product

Catalog No. 12845

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 298.35
Quantity 5mg
Chemical Name or Material Olomoucine
Assay Percent Range >98%
CAS 101622-51-9
Synonym 6-(Benzylamino)-2-(2-hydroxyethylamino)-9-methylpurine
Recommended Storage Desiccate at -20°C
Molecular Formula C15H18N6O

Specifications

Chemical Name or Material Olomoucine
Assay Percent Range >98%
Molecular Formula C15H18N6O
Formula Weight 298.35
Synonym 6-(Benzylamino)-2-(2-hydroxyethylamino)-9-methylpurine
Accessories

Desiccate at -20°C