Please login to your online account to display your discounted pricing

Olomoucine, Tocris

Cyclin-dependent kinase inhibitor

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material Olomoucine
Assay Percent Range >98%
Molecular Formula C15H18N6O
Formula Weight 298.35
Synonym 6-(Benzylamino)-2-(2-hydroxyethylamino)-9-methylpurine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

128425

 
r & d systems
128425
25mg Each for $812.12

12845

 
r & d systems
12845
5mg Each for $192.67
Description & Specifications

Specifications

Formula Weight 298.35
Quantity 25mg
Chemical Name or Material Olomoucine
Assay Percent Range >98%
Molecular Formula C15H18N6O
CAS 101622-51-9
Synonym 6-(Benzylamino)-2-(2-hydroxyethylamino)-9-methylpurine
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C