Please login to your online account to display your discounted pricing

Olomoucine, 98%, high purity, ACROS Organics™

$76.86 - $76.86

Chemical Identifiers

Chemical Name or Material Olomoucine
Name Note 98%
CAS 101622-51-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H18N6O
Formula Weight 298.4
MDL Number MFCD00189360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC328410010

  SDS
thermo fisher scientific
AC328410010
1mg Each for $76.86
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Solid
Formula Weight 298.4
Quantity 1mg
Color White to off-white
Melting Point 120° to 130°C
CAS Max % 100.0
Chemical Name or Material Olomoucine
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C15H18N6O
MDL Number MFCD00189360
CAS 101622-51-9
Synonym 6-Benzylamino-2-(2-hydroxyethylamino)-9-methylpurine
CAS Min % 97.5