missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Olive oil, pure, virgin, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 416540025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Olive oil | |
8001-25-0 | |
0.9135g/mL | |
Authentic | |
1.4670 to 1.4710 | |
184 to 196mg KOH/g | |
14,6836 | |
Pure |
2.0mg KOH/g max. | |
0.2 (270nm, 1g/100ml cyclohexane) max. at 425nm | |
225°C | |
Glass bottle | |
2.5L | |
0.9135 | |
Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C) | |
of fatty acids: 0.1 % max. (< C16) 7.5 to 20.0 % (Palmitic acid) 3.5 % max. (Palmitoleic acid) 0.5 to 5.0 % (Stearic acid) 56.0 to 85.0 % (Oleic acid) 3.5 to 20.0 % (Linoleic acid) 1.2 % max. (Linolenic acid) 0.7 % max. (Arachidic acid) 0.4 % max. (Eic |
Safety and Handling
EINECSNumber : 232-277-
RUO â Research Use Only