Please login to your online account to display your discounted pricing

Alfa Aesar™ Oligomycin A, 99+%

Inhibits growth of molds

Chemical Identifiers

Chemical Name or Material Oligomycin A
CAS 579-13-5
Molecular Formula C45H74O11
Formula Weight 791.07
MDL Number MFCD00065705
Beilstein 5702132
Merck Index 14,6833
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60211LB0

 
alfa aesar™
J60211LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 791.07
Chemical Name or Material Oligomycin A
Physical Form Powder
Assay ≥99%
Molecular Formula C45H74O11
MDL Number MFCD00065705
Beilstein 5702132
Merck Index 14,6833
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Quantity 5mg
Recommended Storage −30° to −10°C
RTECS Number RK3328000
CAS 579-13-5
Solubility Information Soluble in DMSO, ethanol or acetone

Applications
Oligomycin A, is used as inhibitor of mitochondrial ATP synthase and uncoupler of oxidative phosphorylation. Antibiotic, exhibits anti-tumor activity.

Solubility
Soluble in DMSO, ethanol or acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

−30° to −10°C