missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oligodendrocyte Marker Mouse, Clone: O1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen Oligodendrocyte Marker
Clone O1
Host Species Murine
Species Reactivity Chicken, Human, Mouse, Rat
Applications ELISA,Flow Cytometry,Immunocytochemistry,Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013485 Life Technologies
LS14650680
25μg Each for $143.55
5013486 Life Technologies
LS14650682
100μg Each for $361.05
Specifications

Description

This O1 monoclonal antibody reacts with galactocerebroside and other lipids found on late oligodendrocyte precursors. O1 has been reported to react to all species tested (rat, mouse, human, chicken). Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.
Description & Specifications

Specifications

Oligodendrocyte Marker
Murine
ELISA,Flow Cytometry,Immunocytochemistry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
O1
Chicken, Human, Mouse, Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer