missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oleylamine 50.0+%, TCI America™

Manufacturer:  TCI America O005925ML

 View more versions of this product

Catalog No. O005925ML



Specifications

Specifications

Oleylamine
25mL
C18H37N
2735
QGLWBTPVKHMVHM-MDZDMXLPSA-N
(E)-octadec-9-en-1-amine
6258392
Clear Liquid at 20°C
Yellow
112-90-3
MFCD00066507
9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
CCCCCCCCC=CCCCCCCCCN
267.501
267.50
≥50.0% (GC)

Chemical Identifiers

112-90-3
267.501
QGLWBTPVKHMVHM-MDZDMXLPSA-N
6258392
CCCCCCCCC=CCCCCCCCCN
C18H37N
MFCD00066507
9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
(E)-octadec-9-en-1-amine
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0133&(2)-0185&(2)-0176

RTECSNumber : RG4130000

TSCA : Yes

Documents