missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Oleic acid, 99%

Manufacturer:  Alfa Aesar™ 3199714

 View more versions of this product

Catalog No. AA3199714



Specifications

Specifications

Oleic acid
0.887g/mL
C18H34O2
1726542
cis-9-Octadecenoic acid
CCCCCCCCC=CCCCCCCCC(=O)O
1.4610
445639
282.47
286°C (100mmHg)
13°C to 14°C
Air Sensitive
112-80-1
99%
MFCD00064242
14,6828
ZQPPMHVWECSIRJ-KTKRTIGZSA-N
(Z)-octadec-9-enoic acid
282.468
CHEBI:16196
Liquid
Colorless to Yellow
25g

Chemical Identifiers

112-80-1
282.468
ZQPPMHVWECSIRJ-KTKRTIGZSA-N
445639
(Z)-octadec-9-enoic acid
C18H34O2
MFCD00064242
cis-9-Octadecenoic acid
CHEBI:16196
CCCCCCCCC=CCCCCCCCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : RG2275000

TSCA : TSCA

SDS Documents