missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Okadaic acid potassium salt, 98%

Chemical Identifiers

CAS 155751-72-7
Molecular Formula C44H67KO13
Molecular Weight (g/mol) 843.105
MDL Number MFCD00214356
InChI Key UXRQUXBFVICHQJ-UHFFFAOYSA-M
Synonym okadaic acid, potassium salt, okadaic acid potassium salt, okadaic acidpotassium salt
PubChem CID 44134868
IUPAC Name potassium;2-hydroxy-3-[11-hydroxy-2-[4-[4-hydroxy-2-[1-hydroxy-3-(3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl)butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]-4-methyl-1,7-dioxaspiro[5.5]undec-4-en-8-yl]
SMILES CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61791IAM View Documents Alfa Aesar™
J61791IAM
50μg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61791EXG View Documents Alfa Aesar™
J61791EXG
100μg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61791EXS View Documents Alfa Aesar™
J61791EXS
300μg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Okadaic Acid, Potassium Salt has been used as a response modifier in the study of large granular lymphocyte and T-cell biochemistry. In addition to inhibitive activities it is a tumor promoter and has been used to study nitric oxide metabolism, apoptosis, cell cycle and calcium signaling. It is also used as a PP1/PP2A/PP2B/PP2C inhibitor, used to study tumor promotion/NO metabolism/apoptosis

Solubility
Soluble in DMSO or ethanol. Soluble in water at 0.1mg/ml. Also soluble in methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

155751-72-7
843.105
UXRQUXBFVICHQJ-UHFFFAOYSA-M
44134868
CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[K+]
C44H67KO13
MFCD00214356
okadaic acid, potassium salt, okadaic acid potassium salt, okadaic acidpotassium salt
potassium;2-hydroxy-3-[11-hydroxy-2-[4-[4-hydroxy-2-[1-hydroxy-3-(3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl)butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]-4-methyl-1,7-dioxaspiro[5.5]undec-4-en-8-yl]

Specifications

155751-72-7
MFCD00214356
okadaic acid, potassium salt, okadaic acid potassium salt, okadaic acidpotassium salt
UXRQUXBFVICHQJ-UHFFFAOYSA-M
potassium;2-hydroxy-3-[11-hydroxy-2-[4-[4-hydroxy-2-[1-hydroxy-3-(3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl)butyl]-3-methylidenespiro[4a,7,8,8a-tetrahydro-4H-pyrano[3,2-b]pyran-6,5'-oxolane]-2'-yl]but-3-en-2-yl]-4-methyl-1,7-dioxaspiro[5.5]undec-4-en-8-yl]
44134868
Powder
White
Okadaic acid potassium salt
C44H67KO13
UN3462
Soluble in DMSO or ethanol; Soluble in water at 0.1mg/ml; Also soluble in methanol
CC1CCC2(CCCCO2)OC1C(C)CC(C3C(=C)C(C4C(O3)CCC5(O4)CCC(O5)C=CC(C)C6CC(=CC7(O6)C(CCC(O7)CC(C)(C(=O)[O-])O)O)C)O)O.[K+]
843.105
843.11
98%
50μg
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

storageNote1 : -30° to -10°C

Recommended Storage : −30° to −10°C