Parker domnick hunter™ Oil-XPlus™ Filter Elements for Gas Generators
Replacement elements for >Oil-XPlus Compressed Air Filters used with Zero Air Generators
$237.49 - $237.49
Specifications
Certifications/Compliance | ISO 8573-1 |
---|---|
For Use With (Equipment) | Uhp-10za and uhp-35za |
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
14-950-18
|
Parker domnick hunter™
K009AO |
Inlet Filter Element |
Each for $237.49
|
|
|||||
Description
- Use specially treated borosilicate glass microfibers to repel oil and water, reducing pressure drop and minimizing running costs
- Comply with ISO 8573-1 Air Quality Class 1.3.1
- Maximum usage: 8000 hours
Specifications
ISO 8573-1 | |
Uhp-10za and uhp-35za |