missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oil, for heat transfer, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC268770100
Specifications
Oil | |
230°C | |
Plastic drum | |
30 mm2/s (40°C) | |
-6.0°C | |
64742-65- | |
MFCD00147787 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
452.363 | |
Viscous Liquid |
0.8600g/mL | |
Authentic | |
0.86 | |
Yellow | |
10L | |
C16H10N2Na2O7S2 | |
acid orange 10, c.i. acid orange 10, c.i. food orange 4, c.i. orange g, colacid orange g, dolkwal orange g, hexacol orange g, light orange g, orange g, wool orange 2g | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate | |
9566064 |
Chemical Identifiers
64742-65- | |
452.363 | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
9566064 | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
C16H10N2Na2O7S2 | |
MFCD00147787 | |
acid orange 10, c.i. acid orange 10, c.i. food orange 4, c.i. orange g, colacid orange g, dolkwal orange g, hexacol orange g, light orange g, orange g, wool orange 2g | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
Safety and Handling
EINECSNumber : 265-169-7