missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Triple Beam Balances

Accurate, rugged and dependable. OHAUS Triple Beam Balances have stainless-steel weighing pans and bearing covers which prevent rust and corrosion.

Manufacturer:  OHAUS™ 610-00

 View more versions of this product

Catalog No. S400471


 

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.


Specifications

Description

Balance; Triple-beam; Ohaus; 610g capacity; Front beam grad.: 100-500g in 100g div.; Rear beam grad.; 10-100g in 10g div.; Vernier dial grad: 0-10g in 0.1g div.; Dial-O-Gram model; Platform type: Pan with 225g tare beam

  • Precision-cast poises and deep notches in beams for accurate values every time
  • Precision-ground knife edges and self-aligning agate bearings for maximum sensitivity and reproducibility
  • Accurate with quick response time
  • Magnetic damping minimizes oscillation
  • Heavy zinc base won't tip easily like cast aluminum

Specifications
  • Capacity: 610g (2610g with attachment weights)
  • Readability: 0.1g
  • Front Beam: 10 × 0.1g
  • Middle Beam: 500 × 100g
  • Rear Beam: 100 × 10g
  • Vernier Dial (Dial-O-Gram™ Model only): 10 × 0.1g
  • Pan size: 6 in. dia. (21.2cm)
  • Dimensions: 18L × 12W × 9 in.H (45.7 × 30.5 × 22.3cm)

Dial-O-Gram model includes advanced features
  • Direct-reading vernier dial for faster weighing results
  • Tare beam allows containers up to 225g to be balanced out
  • Add rod and clamp accessory (S40022B) to perform specific gravity determinations 
 
Specific Gravity Rod and Clamp
  • Suspends balance above base of table to measure specific gravity.
 
Poly Scoop and Counterweight
  • Ideal for retaining all of the object being weighed
  • Counterweight returns balance to zero when nothing but the scoop is on the platform
  • Stainless-steel scoop and counterweight feature heavy-duty construction for long wear

Note
S40028 features a plastic container

Warranty

Five years

Description & Specifications

Specifications

500 x 100g (middle beam), 100 x 10g (rear beam), 10 x 0.1g (dial)
1.34 lb., 0.61kg
22.86cm
30.48cm
1610-00
5 Years
3.2kg
45.72cm
0.61kg
Triple beam Dial-O-Gram™ balance
1.34 lb.
9 in.
12 in.
Stainless Steel
0.01g
7.05 lb.
18 in.
6 in.
15.2cm