OHAUS™ Starter Water Analysis Bench Meters: Model 3100
Durable, accurate bench meters for a variety of lab applications.
Manufacturer:  OHAUS™ 30058489
 View more versions of this product
Catalog No. S05076
Specifications
Description
Meter; Benchtop; Ohaus; ST3100-F; Water analysis meter with ST310 3-in-1 plastic refillable pH electrode, pH buffer powder sachet and in-use cover; Model Series 3100; Size: 220 × 175 × 78mm; Weight: 0.75kg0
  • Reliable pH testing
  • Intuitive software/organized display for usability
  • Clearly marked input keys
  • Built-in or standalone electrode holders
  • Reliable calibration for accurate measurement
  • Three-point calibration for high accuracy
  • Automatic and manual temperature compensation
  • Automatic and manual endpoint functions show reading stability
  • Height adjustable standalone electrode holder
  • Auto buffer recognition minimizes errors during calibration process
  • 99 memory sets for instant recall of recent calibration data
  • Self-diagnostic software
  • Icon display confirms accuracy of calibration process
  • Connection to peripheral devices through RS232 port
  • Size: 220 × 175 × 78mm
  • Weight: 0.75kg
Description & Specifications
Specifications
ST3100-F
3100 Meter with ST310 3-in-1 Plastic Refillable pH Electrode, pH Buffer Powder Sachet, In-Use Cover, Quick reference guide with operating instructions
0.01 pH; 1 mV; 0.1°C
Backlit LCD