missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Starter Water Analysis Bench Meters: Model 2100

Durable, accurate bench meters for a variety of lab applications. OHAUS Starter Water Analysis Bench Meters: Model 2100 provides reliable pH testing with intuitive software/organized display for usability.

Manufacturer:  OHAUS™ 30057495

 View more versions of this product

Catalog No. 361045916



Includes: Water analysis meter with ST210 2-in-1 plastic refillable pH electrode and pH buffer powder sachet

Description

Description

  • Clearly marked input keys
  • Built-in or standalone electrode holders
  • Reliable calibration for accurate measurement
  • Built-in electrode arm for simple and convenient operation
  • Auto buffer minimizes errors during calibration process
  • Reliable measurements
  • Cost effective
  • Size: 220 x 175 x 78mm
  • Weight: 0.75kg
Specifications

Specifications

Water analysis meter with ST210 2-in-1 plastic refillable pH electrode and pH buffer powder sachet
ST2100-E
0.01 pH; 1 mV; 0.1°C
LCD
SDS
Documents