missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Spring Scales: Dial Type

Designed to stand up to heavy student use and withstand sudden load releases without breaking. OHAUS Dial Type Spring Scales have rugged shatterproof plastic and metal construction.

$35.80 - $76.30

Specifications

Calibration External
Height (Metric) 20.32cm
Material Plastic, Metal
Type Dial spring scale
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Readability Price Quantity  
S32122 OHAUS™
8013-MN
2.2 lb. 1000 x 10g 10g Each for $40.50
S40130 OHAUS™
OHS801100
0.55 lb. 250 x 2g 2g Each for $35.80
S40133 OHAUS™
OHS8012NO
1.1 lb. 500 x 5g (5 x 0.05N) 5g Each for $76.30
S40138 OHAUS™
OHS8014NO
4.4 lb. 2000 x 10g 10g Each for $35.80
Specifications

Description

  • Simple zero adjustment
  • Easy-to-read black graduations on a sturdy frosted-aluminum background
  • Color-coded styles for easy identification of capacity

Order Info

Dial- and pull-type scales are available as well as 8 in. (20.3cm) class demonstration size scales.

Description & Specifications

Specifications

External
20.32cm
Plastic, Metal
Dial spring scale