OHAUS™ Pioneer™ Analytical Balances: Excal Model, 90mm Platform
For basic routine weighing in variety of laboratory, industrial and education applications
$2,483.37 - $3,311.16
Specifications
Material ABS Plastic, Stainless Steel, Glass
Display Type Backlit LCD
Certifications/Compliance OIML E2 or ASTM Class 1
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
02-112-300 OHAUS™
OHS30208441RM
PA124 3 sec. 120g 0.1mg Each for $2,981.23
02-112-298 OHAUS™
OHS30208440RM
PA84 3 sec. 80g 0.1mg Each for $2,483.37
02-112-302 OHAUS™
OHS30208442RM
PA224 3 sec. 220g 0.1mg Each for $3,311.16
Specifications
Description
The Ohaus Pioneer Series of analytical and precision balances are designed for basic routine weighing in variety of laboratory, industrial and education applications. Now with both internal and external calibration models and right combination of performance and features, Ohaus Pioneer offers uncomplicated performance for all basic weighing needs.
  • External calibration (Excal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • RS-232 interface
  • Easy-to-clean analytical draftshield with easy-to-remove-and-replace glass panels, including three sliding doors
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket
Applications
Weighing, parts counting, percent weighing, animal/dynamic weighing and density determination
Compliance
OIML E2 or ASTM Class 1
Description & Specifications
Specifications
ABS Plastic, Stainless Steel, Glass
Backlit LCD
OIML E2 or ASTM Class 1