missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Pioneer™ Precision Balances: Incal Model, 120mm Platform

For basic routine weighing in variety of laboratory, industrial and education applications.

Manufacturer:  OHAUS™ OHS30208459RM

 View more versions of this product

Catalog No. 02-112-297


 

Description

Description

The Ohaus Pioneer Series of analytical and precision balances are designed for basic routine weighing in variety of laboratory, industrial and education applications. Now with both internal and external calibration models and right combination of performance and features, Ohaus Pioneer offers uncomplicated performance for all basic weighing needs.

 

  • Internal calibration (Incal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • RS-232 interface
  • Easy-to-clean analytical draftshield with easy to remove-and-replace glass panels, including three sliding doors
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket

 

Applications
Weighing, parts counting, percent weighing, animal/dynamic weighing and density determination

Compliance

OIML F1 or ASTM Class 2

Specifications

Specifications

120mm
OIML F1 or ASTM Class 2
12.6 in.
1 mg
3 sec.
4.5kg
32 x 19.6 x 28.7cm
28.7cm
PA523C
7.7 in.
Analytical And Precision Balances Incal Model
User selectable
32cm
Backlit LCD
500g
6.9kg
11.3 in.
2mg
1mg
19.6cm
Documents