OHAUS™ Pioneer™ Precision Balances: Incal Model, 120mm Platform
For basic routine weighing in variety of laboratory, industrial and education applications.
$1,894.67 - $2,475.00
Specifications
Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
02-112-293 OHAUS™
OHS30208457RM
PA163C 3 sec. 160g 1mg Each for $1,894.67
02-112-295 OHAUS™
OHS30208458RM
PA323C 3 sec. 320g 1mg Each for $2,143.60
02-112-297 OHAUS™
OHS30208459RM
PA523C 3 sec. 520g 1mg Each for $2,475.00
Specifications
Description
The Ohaus Pioneer Series of analytical and precision balances are designed for basic routine weighing in variety of laboratory, industrial and education applications. Now with both internal and external calibration models and right combination of performance and features, Ohaus Pioneer offers uncomplicated performance for all basic weighing needs.
  • Internal calibration (Incal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • RS-232 interface
  • Easy-to-clean analytical draftshield with easy to remove-and-replace glass panels, including three sliding doors
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket
Applications
Weighing, parts counting, percent weighing, animal/dynamic weighing and density determination
Compliance
OIML F1 or ASTM Class 2
Description & Specifications
Specifications
Backlit LCD
OIML F1 or ASTM Class 2