Please login to your online account to display your discounted pricing

OHAUS™ Pioneer™ Precision Balances: Incal Model, 180mm Platfrom

For basic routine weighing in variety of laboratory, industrial and education applications.

$1,678.14 - $1,906.46

Specifications

Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

02-112-289

 
ohaus™
OHS30208464RM
Each for $1,678.14

02-112-291

 
ohaus™
OHS30208465RM
Each for $1,906.46
Description & Specifications

Specifications

Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2

The Ohaus Pioneer Series of analytical and precision balances are designed for basic routine weighing in variety of laboratory, industrial and education applications. Now with both internal and external calibration models and right combination of performance and features, Ohaus Pioneer offers uncomplicated performance for all basic weighing needs.

  • Internal calibration (Incal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • Easy-to-clean analytical draftshield with easy to remove-and-replace glass panels, including three sliding doors
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket

Applications

  • Weighing
  • Parts counting
  • Percent weighing
  • Animal/dynamic weighing
  • Density determination

 

OIML F1 or ASTM Class 2