OHAUS™ Pioneer™ Precision Balances: Excal Model, 180mm Platform
For basic routine weighing in variety of laboratory, industrial and education applications
$1,338.89 - $2,248.18
Specifications
Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
02-112-280 OHAUS™
OHS30208446RM
PA1602 3 sec. 1.6kg 10mg Each for $1,338.89
02-112-282 OHAUS™
OHS30208447RM
PA2202 3 sec. 2.2kg 10mg Each for $1,573.11
02-112-284 OHAUS™
OHS30208448RM
PA3202 3 sec. 3.2kg 10mg Each for $1,905.98
02-112-286 OHAUS™
OHS30208449RM
PA4202 3 sec. 4.2kg 10mg Each for $2,248.18
Specifications
Description
The Ohaus Pioneer Series of analytical and precision balances are designed for basic routine weighing in variety of laboratory, industrial and education applications. Now with both internal and external calibration models and right combination of performance and features, Ohaus Pioneer offers uncomplicated performance for all basic weighing needs.
  • External calibration (Excal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • RS-232 interface
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket
Applications
Weighing, parts counting, percent weighing, animal/dynamic weighing and density determination
Compliance
OIML F1 or ASTM Class 2
Description & Specifications
Specifications
Backlit LCD
OIML F1 or ASTM Class 2