Please login to your online account to display your discounted pricing

OHAUS™ Pioneer™ Precision Balances: Excal Model, 180mm Platform

For basic routine weighing in variety of laboratory, industrial and education applications

$1,393.88 - $1,620.70

Specifications

Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

02-112-288

 
ohaus™
OHS30208450RM
Each for $1,393.88

02-112-290

 
ohaus™
OHS30208453RM
Each for $1,620.70
Description & Specifications

Specifications

Display Type Backlit LCD
Certifications/Compliance OIML F1 or ASTM Class 2

 

  • External calibration (Excal) model
  • Bright backlit LCD
  • Stainless-steel bottom
  • Easy-to-clean analytical draftshield with easy to remove-and-replace glass panels, including three sliding doors
  • Selectable environmental settings: Three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
  • Design features: Stability Indicator, Auto tare, Manual calibration lock, Software lockout menu, Software reset menu, User selectable communication and printing settings, Auto standby, Mechanical and software overload/underload protection, Up-front level bubble indicator, Intergral weigh below hook, Security bracket

 

Applications

 

  • Weighing
  • Parts counting
  • Percent weighing
  • Animal/dynamic weighing
  • Density determination

 

OIML F1 or ASTM Class 2