OHAUS™ Navigator™ XL Balance
Ultrafast stabilization in less than one second provides fast, accurate and repeatable weighing results
$796.86 - $1,109.60
Specifications
Material ABS Plastic, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232 or USB connectivity
Display Type Backlit LCD
Certifications/Compliance OIML F2 or ASTM CL 4, ISO 9001:2008, NIST
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
01-918-312 OHAUS™
83033062
NVL1101/1 ≤1 sec. 1100g 0.1g Each for $869.04
01-918-311 OHAUS™
83033058
NVL511/1 ≤1 sec. 510g 0.1g Each for $796.86
01-918-313 OHAUS™
83033066
NVL2101/1 ≤1 sec. 2100g 0.1g Each for $942.68
01-918-314 OHAUS™
83033070
NVL5101/1 ≤1 sec. 5100g 0.5g Each for $907.33
01-918-315 OHAUS™
83033074
NVL10000/1 ≤1 sec. 10,000g 1g Each for $1,109.60
01-918-316 OHAUS™
83033078
NVL20000/1 ≤1 sec. 20,000g 1g Each for $1,014.86
Specifications
Description
Two touchless sensors free up your hands for handling samples and limiting keypad wear and tear.
  • Superior mechanical overload protection system and integrated shipping lock
  • Withstands loads up to 400% of its rated capacity
  • Optional RS-232 or USB or ethernet interface kits with integrated cables
  • Up front illuminated level indicator
  • Removable stainless-steel weighing platform
  • Menu lockswitch
  • Integral security bracket and integral shipping lock
  • Stability indicator, overload and underload indicators, low battery indicator, auto shut-off
  • ABS plastic housing; 304 stainless steel pan
Description & Specifications
Specifications
ABS Plastic, Stainless Steel
RS232 or USB connectivity
OIML F2 or ASTM CL 4, ISO 9001:2008, NIST
100/240V 50/60Hz
Backlit LCD