Please login to your online account to display your discounted pricing
OHAUS™ Navigator™ Portable Balances
Touchless sensors that free up students' hands.
$619.65 - $696.60
Specifications
Material ABS Plastic, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232 or USB connectivity
Display Type LCD
Certifications/Compliance cUL
View More Specs
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
S02261
ohaus™
83998182
NVT1601/1 ≤1 sec. 1600g 0.1g Each for $642.60
S02262
ohaus™
83998186EMD
NVT3201/1 ≤1 sec. 3200g 0.2g Each for $643.95
S02263
ohaus™
83998190DUP
NVT6401/1 ≤1 sec. 6400g 0.5g Each for $619.65
S02264
ohaus™
83998194
NVT10001/1 ≤1 sec. 10,000g 0.5g Each for $696.60
S02265
ohaus™
83998198
NVT16000/1 ≤1 sec. 16,000g 1g Each for $696.60
Description & Specifications
Specifications
Material ABS Plastic, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232 or USB connectivity
Display Type LCD
Certifications/Compliance cUL
Achieve faster, accurate, more reliable and repeatable weighing results within less than a second.
Ideal for a wide range of classroom and laboratory weighing applications
  • Highly versatile — a variety of weighing and application modes
  • Two touchless sensors can be independently programmed for up to 36 combinations of operations
Built for longer life
  • Prolong the life of the keypad with touch-free tare, zero, print, function, backlight and OFF operations
  • Superior cleanliness — touchless sensors allow students to focus on handling samples and keeping scale free of contamination
  • Stainless-steel pan, corrosion-free plastic housing and easy-to-clean, spill-resistant pan housing design
  • With superior mechanical overload protection system so scales can withstand loads up to 400% of rated capacity
  • Integrated shipping lock protects the load cell from damage during storage and transport
One year