OHAUS™ Navigator™ NVT Balances
Includes touchless sensors
$856.14 - $856.14
Specifications
Battery Life 100 hr.
Battery Type Internal rechargeable Lead Acid battery
Certifications/Compliance ISO 9001:2008, C Tick, CE Class B, FCC Class B, ICES-003 Class B, 2002/96/EC on WEEE, 2002/95/EC
Connection Type RS232 or USB
Depth (English) 9.8 in.
View More Specs
Includes: 2 Touchless sensors, AC adapter, internal rechargeable battery; front-mounted level bubble with illumination/adjustable leveling feet with skid-resistant Rubber pads, menu lock switch, integral security bracket, integral shipping lock
Products 1
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
01-922-155 View Documents Ohaus™
83998198*
32 lb., 16 kg NVT16000/1 1g Each for $856.14
Description
Description
Combines features, versatility and performance to fit a wide range of industrial, food and laboratory weighing applications.
  • Mechanical overload protection system and integrated shipping lock
  • Two touchless sensors for handling samples and limiting keypad wear and tear
  • Up front illuminated level indicator
  • Removable stainless-steel weighing platform, menu lockswitch, integral security bracket, integral shipping lock, stability indicator, overload and underload indicators, low battery indicator, auto shut-off
  • ABS plastic housing, 304 stainless-steel pan
  • Optional RS232 or USB or ethernet interface kits with integrated cables
  • Liquid crystal display (LCD ) with backlight
  • Application modes include percent weighing, check weighing, parts counting, and accumulation/totalization
Warranty
One year
Specifications
Specifications
100 hr.
ISO 9001:2008, C Tick, CE Class B, FCC Class B, ICES-003 Class B, 2002/96/EC on WEEE, 2002/95/EC
9.8 in.
25cm
2 Touchless sensors, AC adapter, internal rechargeable battery; front-mounted level bubble with illumination/adjustable leveling feet with skid-resistant Rubber pads, menu lock switch, integral security bracket, integral shipping lock
LCD
Gram, kilogram, pound, ounce (decimal or fractional), pound: ounce (decimal or fractional), ounce troy,
7cm
ABS Plastic, Stainless Steel
Internal rechargeable Lead Acid battery
RS232 or USB
6.9 in.
17.4cm
Navigator™ NVT bench scale
100/240V 50/60Hz
2.9 in.
50/60Hz
<1 sec.