Please login to your online account to display your discounted pricing
OHAUS™ Navigator™ NV Balance
Offers a combination of features, versatility and performance that fit a wide range of industrial, food and laboratory weighing applications
$305.91 - $709.37
Specifications
Material ABS Plastic, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232, USB or Ethernet Interface Kits (optional)
Display Type Backlit LCD
Certifications/Compliance cUL
View More Specs
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
01-918-317
ohaus™
83033081
NV212 ≤1 sec. 210g 0.01g Each for $515.39
01-918-319
ohaus™
83033085
NV1101 ≤1 sec. 1100g 0.1g Each for $431.11
01-918-318
ohaus™
83033083
NV511 ≤1 sec. 510g 0.1g Each for $347.47
01-918-321
ohaus™
83033089
NV4101 ≤1 sec. 4100g 0.2g Each for $708.95
01-918-320
ohaus™
83033087
NV2101 ≤1 sec. 2100g 0.1g Each for $557.98
01-918-322
ohaus™
83033091
NV5101 ≤1 sec. 5100g 0.5g Each for $709.37
01-922-156
ohaus™
OHSNV211
NV211 ≤1 sec. 210g 0.1g Each for $305.91
01-922-157
ohaus™
OHSNV4000
NV4000 ≤1 sec. 4000g 1g Each for $321.77
Description & Specifications
Specifications
Material ABS Plastic, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232, USB or Ethernet Interface Kits (optional)
Display Type Backlit LCD
Certifications/Compliance cUL
Two touchless sensors free up your hands for handling samples and limiting keypad wear and tear.
  • Ultra-fast stabilization in less than one second
  • Mechanical overload protection system and integrated shipping lock
  • Up front illuminated level indicator
  • Removable stainless steel weighing platform
  • Menu lockswitch
  • Integral security bracket and shipping lock
  • Stability indicator
  • Overload and underload indicators
  • Low battery indicator and auto shut-off