OHAUS™ Harvard Trip Balances
Ideal for comparative weighing. OHAUS Harvard Trip Balances with choice of single or double beam models provide many years of maintenance-free, comparative weighing using a known standard.
Supplier:  OHAUS™ 80000004
 View more versions of this product
Catalog No. 02-037-SD
Description
Description
  • With 2000g capacities, 0.1g readabilities, and either a stainless-steel 6 in. (15.2cm) diameter x 0.75 in. deep (1.9cm) pan or 6 in. diameter (15.2cm) plate
  • Angled beams with center-reading poises for excellent visibility of results; sliding, rotating poise for fine adjustment
  • Magnetic damping to slow oscillations and speed weighings
  • Zero-adjust compensator ensures precision and reproducibility
  • Equipped for specific gravity weighing
  • Precision-ground knives, covered self-aligning bearings, cross-braced aluminum-alloy box-end beam, and elliptical check pins
  • Single-beam balances graduated in 0.1g increments; double-beams in 0.1 and 10g increments
Specifications
Specifications
6 in.
7 in.
5.9 in.
15 cm
24.1 cm
Stainless Steel
35.5 cm
4.4 lb., 2 kg
15.2 cm
10 x 0.1g (top beam), 200 x 10g (bottom beam)
22.8 cm
9.5 in.
1550-SD
14 in.
5 Years
Harvard Double Beam Balance