OHAUS™ FD Food Portioning Scales
Speed, durability and cleanliness. OHAUS FD Food Portioning Scales are ideally suited for food preparation in restaurants, bakeries, food manufacturing and other areas requiring fast operation and flexibility.
$1,413.21 - $1,579.02
Specifications
Automation Interface 4 Button membrane switch
Battery Life 120 Hr.
Battery Type Internal rechargeable lead acid battery
Calibration User selectable external span or linearity calibration
Certifications/Compliance ISO 9001:2008, NSF/ANSI 169-2005, USDA/AMS, HACCP, cULus, AS/NZS4251.1, Emission, AS/NZS4252.1, Immunity, FCC Class B Part 15 operation, NTEP Class 3 to NIST Handbook 44, OIML R76 Class 3
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
02-113-29
View Documents
OHAUS™
83998148
6 kg FD6H 0.0005 kg
Each for $1,579.02
02-113-30
View Documents
OHAUS™
83998149
13.6 kg FD15H 0.0005 kg
Each for $1,501.63
0211327
View Documents
OHAUS™
83998119
13.6 kg, 30 lb. FD15 0.002 kg
Each for $1,413.21
0211325
View Documents
OHAUS™
83998117
2.72 kg, 6 lb. FD3 0.0005 kg
Each for $1,413.21
Description
Description
Standard Features
  • Rugged stainless-steel construction
  • Removable, dishwasher-safe, stainless-steel weighing pan
  • Simple 4-key operation and plain-text menu for fast setup
  • 3/4 in. high backlit LCD display with text annunciators Fast display update (under 2 seconds); large, dedicated tare key Auto tare for hands-free operation
  • Optional foot tare switch for hands-free operation
  • Checkweighing mode included with advanced features
  • 3-LED indication of Over, Accept and Under with audible buzzer
  • Positive mode for packaging, negative checkweighing mode for portioning, and “zero” checkweighing mode for sorting and grading operations
  • Dedicated Target key for entry of setpoints for checkweighing mode
Advanced Features
  • Selectable environmental averaging settings Backlight control with auto-off option Selectable weighing units Configurable checkweighing mode audible buzzer Configurable software lockout menu Span or linearity calibration in kg or lb. unit with optional external masses Selectable auto-zero tracking
  • NTEP - Legal for Trade certified
  • Low-battery detection system
  • Adjustable feet with integrated level bubble
Service Plan
Service agreement available.
Specifications
Specifications
4 Button membrane switch
Internal rechargeable lead acid battery
ISO 9001:2008, NSF/ANSI 169-2005, USDA/AMS, HACCP, cULus, AS/NZS4251.1, Emission, AS/NZS4252.1, Immunity, FCC Class B Part 15 operation, NTEP Class 3 to NIST Handbook 44, OIML R76 Class 3
7.1 cm
Rubber pan supports, Stainless Steel cover
LCD
Pound, decimal ounce, fractional ounce, gram, kilogram
71 mm
Stainless Steel
14°F to 104°F
120 Hr.
User selectable external span or linearity calibration
8.2 in.
20.9 cm
FD Food Portioning Balance
100/120 V 50/60 Hz
2.8 in.
50/60 Hz
<2 sec.
-10°C to +40°C
SDS