OHAUS™ Defender™ 7000 Bench Scales
Offers multiple features, durability, and easy configuration. Ideal for shipping and receiving, production, packaging and general commercial and industrial weighing applications.
$1,780.79 - $2,938.98
Specifications
Automation Interface Positive feedback membrane keys, 5 function keys and alpha-numeric keypad
Battery Life 80 hr.
Battery Type 6 C
Calibration Span 1kg or 1 lb. to 100% of capacity/linearity: 3 or 5 point
Certifications/Compliance ISO 9001:2008, NTEP (CC No. 09-102 for indicator, 01-048A1 for bases), (AM-5762 for Indicator, AM-5719 for bases), cULus 2005/95/EC low voltage directive: EN60950-1:2001
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Model Readability Price Quantity  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Model Readability Price Quantity  
01919805 OHAUS™
80501477
25 lb. 10000g D71P10QR1 0.0001kg Each for $1,780.79
01919806 OHAUS™
80501478
50 lb. 25000g D71P25QR1 0.002kg Each for $1,875.53
01919807 OHAUS™
80501479
100 lb. 50000g D71P50QL2 0.005kg Each for $1,979.63
01919808 OHAUS™
80501480
250 lb. 100000g D71P100QL2 0.01kg Each for $2,074.37
01919809 OHAUS™
80501481
500 lb. 250000g D71P250QX2 0.02kg Each for $2,938.98
Specifications
Description
Ideal for shipping and receiving, production, packaging and general commercial and industrial weighing applications.
  • Full alphanumeric keypad, dual-line display supporting English, Spanish or French text, checkweighing LEDs
  • Advanced parts counting with dual-scale counting, dynamic weighing, percent weighing, display hold, checkweighing, accumulation, time/date
  • Temporary display of 10x readability in weighing mode using quick key press
  • 255 item library data storage function
  • GMP output with date time, statistics, lb., oz., lb.:oz., g, kg, metric tonnes, user-defined custom unit
  • 304 grade stainless-steel platform pan with black polyurethane painted steel form-and weld frame, IP67 aluminium load cell, stainless-steel column and wall bracket included
  • LCD display with white LED backlight, alphanumeric line and multi-language, plus 3 color checkweigh LED
  • RS232 interface with GMP, Time-Date, optional RS422/485, AC/DC Relay, 2nd RS232 interface
  • Internal universal power supply, 6 x C batteries or optional internal NiMH rechargeable battery
Description & Specifications
Specifications
Positive feedback membrane keys, 5 function keys and alpha-numeric keypad
6 C
ISO 9001:2008, NTEP (CC No. 09-102 for indicator, 01-048A1 for bases), (AM-5762 for Indicator, AM-5719 for bases), cULus 2005/95/EC low voltage directive: EN60950-1:2001
Painted Steel column
LCD
Gram, kilogram, ounce, pound, metric tones, custom
67
<2 sec.
-10 to 40°C
80 hr.
Span 1kg or 1 lb. to 100% of capacity/linearity: 3 or 5 point
RS232, external input
Defender™ 7000 bench scale
100/240V 50/60Hz
50/60Hz
ABS Plastic, Stainless Steel, Aluminum
14 to 104°F
100/240V