missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Conductivity Standards

Measure the ability of an electrolyte solution to conduct electricity.

$57.28 - $57.28

Specifications

Boiling Point ∽100°C
Capacity 8.45 oz., 250mL
Color Colorless, Clear
pH Range ∽6
Type Conductivity Standards
View More Specs

Products 1
Catalog Number Mfr. No. Conductivity Price Quantity  
Catalog Number Mfr. No. Conductivity Price Quantity  
S05115 View Documents OHAUS™
30065088
1413μS/cm Each for $57.28
 
Description

Description

  • 250mL Solution
  • Available in values of 84μs/cm, 1413μs/cm and 12.88ms/cm

Extremes of temperature and direct sunlight should be avoided.

Specifications

Specifications

∽100°C
Colorless, Clear
Conductivity Standards
Completely Miscible
8.45 oz., 250mL
∽6
Conductivity sensor