Please login to your online account to display your discounted pricing
OHAUS™ CL Portable Balances
Perform basic weighing with a simple two-button keypad. The CL Portable balance is designed with a large weighing platform and can adequately handle large samples including oversized letters.
$121.89 - $184.56
Specifications
Material ABS Plastic
Display Type LCD
Certifications/Compliance OIML M1 or ASTM CL F
View More Specs
Includes:  3 AA Batteries (Included); AC Adapter (Not included)
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
01-919-234
ohaus™
80010610
CL201 3 sec. 200g 0.1g Each for $121.89
01-919-236
ohaus™
80010612
CL5000 3 sec. 5000g 1g Each for $184.56
01-919-235
ohaus™
80010611
CL2000 3 sec. 2000g 1g Each for $121.89
Description & Specifications
Specifications
Material ABS Plastic
Display Type LCD
Certifications/Compliance OIML M1 or ASTM CL F
  • ABS housing, ABS pan
  • Low battery indicator
  • Auto shut-off feature
  • Mechanical and software overload protection
  • External push-button calibration
  • Liquid Crystal Display (LCD)
  • RF protection for use near cell phones and production areas
  • Rugged housing with integral platform ring for stacking and storage