OHAUS™ Bench Scales Computer Data Transfer Cables
Connects Link Champ General-Purpose Bench Scales to IBM-PC computers via standard RS-232-C.
Manufacturer: OHAUS™ 80500433
Description
Permits data collection via computer.
Specifications
Trooper, Trooper Count, CD Indicators | |
RS-232 cable, PC 9 pin | |
Computer data transfer |