missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ODQ, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 328800500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
ODQ | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00792620 | |
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq | |
LZMHWZHOZLVYDL-UHFFFAOYSA-N | |
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one | |
1456 | |
98% |
41443-28-1 | |
100.0 | |
97.5% min. (HPLC) | |
C9H5N3O2 | |
50mg | |
Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,ethylacetate,dmso | |
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23 | |
187.2 | |
187.2 |
Chemical Identifiers
41443-28-1 | |
187.2 | |
LZMHWZHOZLVYDL-UHFFFAOYSA-N | |
1456 | |
C1=CC=C2C(=C1)N=CC3=NOC(=O)N23 |
C9H5N3O2 | |
MFCD00792620 | |
1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq | |
[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one |
RUO â Research Use Only