Please login to your online account to display your discounted pricing

Octyltrichlorosilane, 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC216505000

 View more versions of this product

Catalog No. AC216505000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 247.67
Physical Form Liquid
Quantity 500g
Color Clear colorless to light yellow
Density 1.07g/mL
Beilstein 04, III, 1907
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
CAS 5283-66-9
Assay Percent Range 97%
Molecular Formula C8H17Cl3Si
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
MDL Number MFCD00000488
Linear Formula CH3(CH2)7SiCl3
Infrared Spectrum Authentic
Chemical Name or Material Octyltrichlorosilane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Octyltrichlorosilane
CAS 5283-66-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H17Cl3Si
Linear Formula CH3(CH2)7SiCl3
Formula Weight 247.67
MDL Number MFCD00000488
Accessories

GHS Signal Word: Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

SDS & Safety Documents