Learn More
Octylmethyldichlorosilane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 255260050
Specifications
Octylmethyldichlorosilane | |
95°C | |
Glass bottle | |
0.97 | |
Colorless to Yellow | |
98% | |
96.0 | |
97.5% min. (GC) | |
C8H17Si(CH3)Cl2 | |
acmc-209czn, dichloro methyl octyl silane, dichloro methyl octylsilane, dichloro methyl octylsilane #, dichloro methyl-n-octylsilane, methyloctyl dichlorosilane, methyloctyldichlorosilane, n-octylmethyldichlorosilane, octylmethyldichlorosilane, silane, dichloromethyloctyl | |
QHBMMABVNRSRHW-UHFFFAOYSA-N | |
dichloro-methyl-octylsilane | |
84649 | |
Liquid |
0.9700g/mL | |
Authentic | |
1.442 | |
230°C | |
5mL | |
14799-93-0 | |
100.0 | |
C9H20Cl2Si | |
MFCD00042606 | |
Solubility in water: .. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic(chlorinated) hydrocarbons | |
CCCCCCCC[Si](C)(Cl)Cl | |
227.24 | |
227.24 | |
97% |
Chemical Identifiers
14799-93-0 | |
227.24 | |
QHBMMABVNRSRHW-UHFFFAOYSA-N | |
84649 | |
CCCCCCCC[Si](C)(Cl)Cl |
C9H20Cl2Si | |
MFCD00042606 | |
acmc-209czn, dichloro methyl octyl silane, dichloro methyl octylsilane, dichloro methyl octylsilane #, dichloro methyl-n-octylsilane, methyloctyl dichlorosilane, methyloctyldichlorosilane, n-octylmethyldichlorosilane, octylmethyldichlorosilane, silane, dichloromethyloctyl | |
dichloro-methyl-octylsilane |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 238-863-2
TSCA : TSCA