missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octyl isocyanate 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 416460100

 View more versions of this product

Catalog No. AC416460100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Octyl isocyanate
3158-26-7
100.0
Glass bottle
≥98.5%
1.4290 to 1.4310 (20°C, 589nm)
CH3(CH2)7NCO
MFCD00013887
10g
99%
98.5
Authentic
Liquid
99%
C9H17NO
155.24
Turbid milky to light yellow

Chemical Identifiers

3158-26-7
100.0
C9H17NO
155.24
Octyl isocyanate, 99%
98.5
99%
CH3(CH2)7NCO
MFCD00013887
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-598-1

TSCA : TSCA