Please login to your online account to display your discounted pricing

Octyl chloroformate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314550250

 View more versions of this product

Catalog No. AC314550250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 192.69
Quantity 25g
Color Clear colorless
Density 0.984g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Octyl chloroformate
Molecular Formula C9H17ClO2
CAS 7452-59-7
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00000652
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4300 to 1.4320 (20°C, 589nm)
Linear Formula ClCO(CH)7CH3
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Octyl chloroformate
CAS 7452-59-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H17ClO2
Linear Formula ClCO(CH)7CH3
Formula Weight 192.69
MDL Number MFCD00000652
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents