missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octyl chloroformate, 97%, ACROS Organics™

Chemical Identifiers

CAS 7452-59-7
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H17ClO2
Linear Formula ClCO(CH)7CH3
Formula Weight 192.69
MDL Number MFCD00000652
Chemical Name or Material Octyl chloroformate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC314550250 SDS Thermo Fisher Scientific
AC314550250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC314550050 SDS Thermo Fisher Scientific
314550050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Coupling reagent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

7452-59-7
100.0
C9H17ClO2
192.69
Octyl chloroformate
96.0
97%
ClCO(CH)7CH3
MFCD00000652
≥96.0%

Specifications

7452-59-7
100.0
Authentic
Liquid
1.4300 to 1.4320 (20°C, 589nm)
ClCO(CH)7CH3
MFCD00000652
25g
≥96.0%
96.0
0.984g/mL
Glass bottle
97%
C9H17ClO2
192.69
Clear colorless
Octyl chloroformate

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger