missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octyl chloroformate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 314550050

 View more versions of this product

Catalog No. AC314550050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
192.69
Clear colorless
96.0
Glass bottle
C9H17ClO2
7452-59-7
Authentic
ClCO(CH)7CH3
Liquid
5g
0.984g/mL
100.0
97%
Octyl chloroformate
MFCD00000652
1.4300 to 1.4320 (20°C, 589nm)

Chemical Identifiers

7452-59-7
100.0
C9H17ClO2
192.69
Octyl chloroformate
96.0
97%
ClCO(CH)7CH3
MFCD00000652
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger