Please login to your online account to display your discounted pricing

Octyl acetate, +99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC310635000

 View more versions of this product

Catalog No. AC310635000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 172.27
Assay ≥99.0%
Quantity 500g
Color Clear colorless
TSCA TSCA
RTECS Number AJ1400000
EINECS Number 203-939-6
CAS Min % 99.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4175 to 1.4195 (20°C, 589nm)
Infrared Spectrum Authentic
Water 0.3% max. (K.F.)
Health Hazard 1
MDL Number MFCD00009562
Name Note 99+%
Linear Formula CH3CO2(CH2)7CH3
Molecular Formula C10H20O2
CAS 112-14-1
Chemical Name or Material Octyl acetate
Assay Percent Range 99+%

Specifications

Chemical Name or Material Octyl acetate
Name Note 99+%
CAS 112-14-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H20O2
Linear Formula CH3CO2(CH2)7CH3
Formula Weight 172.27
MDL Number MFCD00009562
Accessories

SDS & Safety Documents